Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCTTTTTTTTCTGGCTAAAATTA[C/T]GTTACTTCCTTTGGTCATCCTGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
22 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609209 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IVNS1ABP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IVNS1ABP - influenza virus NS1A binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006469.4 | 5376 | Intron | NP_006460.2 | |||
XM_005244843.3 | 5376 | UTR 3 | XP_005244900.1 | |||
XM_011509079.2 | 5376 | UTR 3 | XP_011507381.1 | |||
XM_011509080.2 | 5376 | UTR 3 | XP_011507382.1 | |||
XM_017000084.1 | 5376 | UTR 3 | XP_016855573.1 |
SWT1 - SWT1, RNA endoribonuclease homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |