Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGTCTTCCCCAGGCCCTCCACAA[A/G]TCTTAGACATCTAGATCAGAAATTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609562 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPA6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CPA6 - carboxypeptidase A6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020361.4 | Intron | NP_065094.3 | ||||
XM_011517570.2 | Intron | XP_011515872.1 | ||||
XM_017013646.1 | Intron | XP_016869135.1 | ||||
XM_017013647.1 | Intron | XP_016869136.1 |
LOC102724708 - uncharacterized LOC102724708 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |