Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604517 | ||||||||||||||||||||
Literature Links: |
PPARGC1A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPARGC1A - PPARG coactivator 1 alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013261.3 | 2725 | UTR 3 | NP_037393.1 | |||
XM_005248131.4 | 2725 | UTR 3 | XP_005248188.1 | |||
XM_005248132.1 | 2725 | UTR 3 | XP_005248189.1 | |||
XM_005248134.4 | 2725 | UTR 3 | XP_005248191.1 | |||
XM_011513765.2 | 2725 | UTR 3 | XP_011512067.1 | |||
XM_011513766.1 | 2725 | UTR 3 | XP_011512068.1 | |||
XM_011513767.2 | 2725 | UTR 3 | XP_011512069.1 | |||
XM_011513768.1 | 2725 | UTR 3 | XP_011512070.1 | |||
XM_011513769.2 | 2725 | Intron | XP_011512071.1 | |||
XM_011513770.2 | 2725 | UTR 3 | XP_011512072.1 | |||
XM_011513771.1 | 2725 | UTR 3 | XP_011512073.1 | |||
XM_017007664.1 | 2725 | UTR 3 | XP_016863153.1 |