Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTACAGTTCCCTGACATCTCATAA[C/T]CTGAATGTAGCAGCAAATAGCAGTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604732 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TFEC PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TFEC - transcription factor EC | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018058.2 | 4000 | UTR 3 | NP_001018068.1 | |||
NM_001244583.1 | 4000 | UTR 3 | NP_001231512.1 | |||
NM_012252.3 | 4000 | UTR 3 | NP_036384.1 | |||
XM_011515963.1 | 4000 | UTR 3 | XP_011514265.1 | |||
XM_011515964.2 | 4000 | UTR 3 | XP_011514266.1 | |||
XM_011515965.2 | 4000 | UTR 3 | XP_011514267.1 | |||
XM_011515966.2 | 4000 | UTR 3 | XP_011514268.1 | |||
XM_011515969.1 | 4000 | UTR 3 | XP_011514271.1 | |||
XM_011515970.1 | 4000 | UTR 3 | XP_011514272.1 | |||
XM_011515972.2 | 4000 | UTR 3 | XP_011514274.1 | |||
XM_017011874.1 | 4000 | UTR 3 | XP_016867363.1 | |||
XM_017011875.1 | 4000 | UTR 3 | XP_016867364.1 | |||
XM_017011876.1 | 4000 | UTR 3 | XP_016867365.1 | |||
XM_017011877.1 | 4000 | UTR 3 | XP_016867366.1 | |||
XM_017011878.1 | 4000 | UTR 3 | XP_016867367.1 |