Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATAGTCTTTTTCTTACAGCGAAT[A/G]CTAAACTGCTATTGAAATAATGCTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF800 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF800 - zinc finger protein 800 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_176814.4 | Intron | NP_789784.2 | ||||
XM_005250181.3 | Intron | XP_005250238.1 | ||||
XM_006715867.3 | Intron | XP_006715930.1 | ||||
XM_006715868.3 | Intron | XP_006715931.1 | ||||
XM_006715869.3 | Intron | XP_006715932.1 | ||||
XM_011515859.2 | Intron | XP_011514161.1 |
Set Membership: |
HapMap |