Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGGGCCTCCATGTTACATGTGCT[A/G]TGAATGCCAGTTATTAATAAACAAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STRIP2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
STRIP2 - striatin interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134336.1 | Intron | NP_001127808.1 | ||||
NM_020704.2 | Intron | NP_065755.1 | ||||
XM_006716069.3 | Intron | XP_006716132.1 | ||||
XM_011516432.2 | Intron | XP_011514734.1 | ||||
XM_011516435.1 | Intron | XP_011514737.1 | ||||
XM_017012470.1 | Intron | XP_016867959.1 | ||||
XM_017012471.1 | Intron | XP_016867960.1 | ||||
XM_017012472.1 | Intron | XP_016867961.1 |
Set Membership: |
HapMap |