Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGCTCCGCCGCCCAGCCTGACTG[A/G]CTGCATTGGCACGGTGGGCATTTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601497 MIM: 610900 | ||||||||||||||||||||
Literature Links: |
BAG1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BAG1 - BCL2 associated athanogene 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172415.1 | 256 | Intron | NP_001165886.1 | |||
NM_004323.5 | 256 | Intron | NP_004314.5 |
CHMP5 - charged multivesicular body protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195536.1 | 256 | Missense Mutation | GAC,GGC | D,G 19 | NP_001182465.1 | |
NM_016410.5 | 256 | Missense Mutation | GAC,GGC | D,G 19 | NP_057494.3 |