Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGAGGTGAGGACAAAGTCTAGAA[A/C]CCAACAACCCAGCCAGATGCTCTGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601497 MIM: 613929 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BAG1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BAG1 - BCL2 associated athanogene 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172415.1 | 1668 | UTR 3 | NP_001165886.1 | |||
NM_004323.5 | 1668 | UTR 3 | NP_004314.5 |
SPINK4 - serine peptidase inhibitor, Kazal type 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |