Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTTATAGGCTATTATTTACCTTA[C/G]AATATGAAAGTTTAAAGGTCAAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614398 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA1462 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
KIAA1462 - KIAA1462 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020848.2 | 6759 | UTR 3 | NP_065899.1 | |||
XM_011519608.1 | 6759 | UTR 3 | XP_011517910.1 | |||
XM_011519609.2 | 6759 | UTR 3 | XP_011517911.1 |