Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGTTAAACGATGATCATGTTGCT[A/G]TAAAAGACCTTCAAATTTTCATTAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM196 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TMEM196 - transmembrane protein 196 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152774.3 | 2331 | UTR 3 | NP_689987.3 | |||
XM_005249692.4 | 2331 | Intron | XP_005249749.2 | |||
XM_011515249.2 | 2331 | Intron | XP_011513551.1 | |||
XM_011515250.2 | 2331 | Intron | XP_011513552.1 | |||
XM_017011928.1 | 2331 | Intron | XP_016867417.1 | |||
XM_017011929.1 | 2331 | Intron | XP_016867418.1 |