Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAGAAAATAGTCTCTGCCTTAGA[G/T]AATTTATAATTCAATCATCAGTAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610531 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM126A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM126A - family with sequence similarity 126 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032581.3 | Intron | NP_115970.2 | ||||
XM_005249894.3 | Intron | XP_005249951.1 | ||||
XM_005249895.2 | Intron | XP_005249952.1 | ||||
XM_011515589.2 | Intron | XP_011513891.1 | ||||
XM_011515590.1 | Intron | XP_011513892.1 |