Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGATAAGTTGGTGGTTTCCATGGA[G/T]ACTCTTCAAGGCAGGATGGATAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
KBTBD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KBTBD2 - kelch repeat and BTB domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015483.2 | 2440 | Missense Mutation | NP_056298.2 | |||
XM_005249696.2 | 2440 | Missense Mutation | XP_005249753.1 | |||
XM_005249697.2 | 2440 | Missense Mutation | XP_005249754.1 | |||
XM_011515256.2 | 2440 | Missense Mutation | XP_011513558.1 | |||
XM_011515257.2 | 2440 | Missense Mutation | XP_011513559.1 |