Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTGGAGAGTTCAGTATGATTGAC[A/C]GTAAGATACATGGGCAAACTACATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613892 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPY19L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DPY19L1 - dpy-19 like 1 (C. elegans) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015283.1 | Intron | NP_056098.1 | ||||
XM_011515245.2 | Intron | XP_011513547.1 | ||||
XM_011515246.2 | Intron | XP_011513548.1 |