Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTTTTTTTTTTCCTGTTCTGAG[A/T]GCTGTTCCTAGATGGCTTTGGGTTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609062 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
POU6F2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
POU6F2 - POU class 6 homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166018.1 | Intron | NP_001159490.1 | ||||
NM_007252.3 | Intron | NP_009183.3 | ||||
XM_011515113.2 | Intron | XP_011513415.1 |
POU6F2-AS2 - POU6F2 antisense RNA 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |