Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGGTGTCACATATCATCAGAAAC[A/G]CAAGGCTGGGGCACCTTCTGAGCAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609279 MIM: 605793 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CENPJ PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CENPJ - centromere protein J | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018451.4 | Intron | NP_060921.3 | ||||
XM_011535149.2 | Intron | XP_011533451.1 | ||||
XM_011535150.2 | Intron | XP_011533452.1 | ||||
XM_017020673.1 | Intron | XP_016876162.1 |
RNF17 - ring finger protein 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184993.1 | Intron | NP_001171922.1 | ||||
NM_031277.2 | Intron | NP_112567.2 | ||||
XM_006719846.3 | Intron | XP_006719909.1 | ||||
XM_006719849.2 | Intron | XP_006719912.1 | ||||
XM_006719852.2 | Intron | XP_006719915.1 | ||||
XM_011535152.2 | Intron | XP_011533454.1 | ||||
XM_011535155.2 | Intron | XP_011533457.1 | ||||
XM_011535156.2 | Intron | XP_011533458.1 | ||||
XM_011535157.2 | Intron | XP_011533459.1 | ||||
XM_011535158.2 | Intron | XP_011533460.1 | ||||
XM_011535159.2 | Intron | XP_011533461.1 | ||||
XM_011535160.2 | Intron | XP_011533462.1 | ||||
XM_011535162.1 | Intron | XP_011533464.1 | ||||
XM_011535163.1 | Intron | XP_011533465.1 | ||||
XM_011535164.2 | Intron | XP_011533466.1 | ||||
XM_017020676.1 | Intron | XP_016876165.1 |
Set Membership: |
HapMap |