Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTCACCTCAAGGCCTTCTTCAC[C/T]ATCCTGGTTTCCCCAGCTTTGACCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600763 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNORA31 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SNORA31 - small nucleolar RNA, H/ACA box 31 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TPT1 - tumor protein, translationally-controlled 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286272.1 | 2773 | UTR 3 | NP_001273201.1 | |||
NM_001286273.1 | 2773 | UTR 3 | NP_001273202.1 | |||
NM_003295.3 | 2773 | UTR 3 | NP_003286.1 |
TPT1-AS1 - TPT1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |