Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTGAATTTTAGTAGTAAGTATA[A/G]GAAGTTCATTATACTGTCCTCACCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605390 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LPXN PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LPXN - leupaxin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZFP91 - ZFP91 zinc finger protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197051.1 | Intron | NP_001183980.1 | ||||
NM_053023.4 | Intron | NP_444251.1 |
ZFP91-CNTF - ZFP91-CNTF readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |