Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAATTGCATCTTTGCCAGAAACTC[C/T]TAGCAAAAACAAGGCCCAATTTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600876 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STX3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
STX3 - syntaxin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178040.1 | Intron | NP_001171511.1 | ||||
NM_004177.4 | Intron | NP_004168.1 | ||||
XM_005274195.3 | Intron | XP_005274252.1 | ||||
XM_005274198.2 | Intron | XP_005274255.1 | ||||
XM_005274200.3 | Intron | XP_005274257.1 | ||||
XM_011545221.2 | Intron | XP_011543523.1 | ||||
XM_017018188.1 | Intron | XP_016873677.1 | ||||
XM_017018189.1 | Intron | XP_016873678.1 | ||||
XM_017018190.1 | Intron | XP_016873679.1 | ||||
XM_017018191.1 | Intron | XP_016873680.1 | ||||
XM_017018192.1 | Intron | XP_016873681.1 | ||||
XM_017018193.1 | Intron | XP_016873682.1 | ||||
XM_017018194.1 | Intron | XP_016873683.1 | ||||
XM_017018195.1 | Intron | XP_016873684.1 | ||||
XM_017018196.1 | Intron | XP_016873685.1 |
Set Membership: |
HapMap |