Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTAGCTAATCTGGAGGAAAAAGA[C/G]TAAAAAGCACTGAATTTAGCAGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603583 MIM: 615533 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DLG2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DLG2 - discs large MAGUK scaffold protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM126B - transmembrane protein 126B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193537.2 | Intron | NP_001180466.1 | ||||
NM_001193538.2 | Intron | NP_001180467.1 | ||||
NM_001256546.1 | Intron | NP_001243475.1 | ||||
NM_001256547.1 | Intron | NP_001243476.1 | ||||
NM_018480.4 | Intron | NP_060950.3 | ||||
XM_011545165.2 | Intron | XP_011543467.1 |