Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAAAGTACTGTCCCCTTTGAATC[A/G]CGCCTATTGCTGACCCGCAGGTGTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607294 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DLGAP4-AS1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DLGAP4-AS1 - DLGAP4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TGIF2 - TGFB induced factor homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199513.1 | Intron | NP_001186442.1 | ||||
NM_001199514.1 | Intron | NP_001186443.1 | ||||
NM_001199515.1 | Intron | NP_001186444.1 | ||||
NM_021809.6 | Intron | NP_068581.1 |
TGIF2-C20orf24 - TGIF2-C20orf24 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199535.1 | Intron | NP_001186464.1 |
Set Membership: |
HapMap |