Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCATGATAACAAAGAATAAAGCAA[C/T]TGAAGAATTAAGGAAATAGAGTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615215 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNU1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNU1 - potassium calcium-activated channel subfamily U member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031836.2 | Intron | NP_001027006.2 | ||||
XM_011544424.1 | Intron | XP_011542726.1 | ||||
XM_011544426.1 | Intron | XP_011542728.1 | ||||
XM_011544427.1 | Intron | XP_011542729.1 | ||||
XM_017013144.1 | Intron | XP_016868633.1 |