Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCTCCAGGGTCATCAGGTCCTGT[A/G]TTTATAGGACAGAGACACCAGGAGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613919 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KIF6 - kinesin family member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289020.1 | 2039 | UTR 3 | NP_001275949.1 | |||
NM_001289021.1 | 2039 | UTR 3 | NP_001275950.1 | |||
NM_001289024.1 | 2039 | UTR 3 | NP_001275953.1 | |||
NM_145027.4 | 2039 | UTR 3 | NP_659464.3 | |||
XM_005248904.4 | 2039 | Intron | XP_005248961.1 | |||
XM_011514357.2 | 2039 | Intron | XP_011512659.1 | |||
XM_011514358.2 | 2039 | Intron | XP_011512660.1 | |||
XM_011514359.2 | 2039 | Intron | XP_011512661.1 | |||
XM_011514361.2 | 2039 | Intron | XP_011512663.1 | |||
XM_017010427.1 | 2039 | Intron | XP_016865916.1 | |||
XM_017010428.1 | 2039 | Intron | XP_016865917.1 |
Set Membership: |
HapMap |