Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCTGTCTAATCTCAGAAAATTAT[A/G]TAAAATTGTATTAGAAAAAAATGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 112265 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BMP5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BMP5 - bone morphogenetic protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005249304.3 | 2251 | UTR 3 | XP_005249361.1 | |||
XM_011514816.2 | 2251 | Intron | XP_011513118.1 | |||
XM_011514817.2 | 2251 | Intron | XP_011513119.1 | |||
XM_017011198.1 | 2251 | UTR 3 | XP_016866687.1 |
Set Membership: |
HapMap |