Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTAGCACCTTTCTTTTAGGTGACG[A/G]GAAAGATCTCTGTAAATACTGCTGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142910 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMGCR PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HMGCR - 3-hydroxy-3-methylglutaryl-CoA reductase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000859.2 | Intron | NP_000850.1 | ||||
NM_001130996.1 | Intron | NP_001124468.1 | ||||
XM_011543357.1 | Intron | XP_011541659.1 | ||||
XM_011543358.1 | Intron | XP_011541660.1 | ||||
XM_011543359.1 | Intron | XP_011541661.1 |
Set Membership: |
HapMap |