Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGAGTTTTCAGCCTTTTGGTCC[A/C]AAATAGTGCCTATATTCAAATTGTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MBLAC2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MBLAC2 - metallo-beta-lactamase domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLR3G - RNA polymerase III subunit G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006467.2 | Intron | NP_006458.2 | ||||
XM_005248404.3 | Intron | XP_005248461.1 | ||||
XM_011543100.2 | Intron | XP_011541402.1 | ||||
XM_011543101.2 | Intron | XP_011541403.1 | ||||
XM_011543102.2 | Intron | XP_011541404.1 | ||||
XM_011543103.2 | Intron | XP_011541405.1 | ||||
XM_011543104.2 | Intron | XP_011541406.1 | ||||
XM_011543105.2 | Intron | XP_011541407.1 | ||||
XM_017008957.1 | Intron | XP_016864446.1 |
Set Membership: |
HapMap |