Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAACTTGCTATACGAAGGACAAAT[A/G]TATGTATACAAACAAATGCACACGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605250 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCC4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ABCC4 - ATP binding cassette subfamily C member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105515.2 | 5494 | Intron | NP_001098985.1 | |||
NM_001301829.1 | 5494 | UTR 3 | NP_001288758.1 | |||
NM_001301830.1 | 5494 | Intron | NP_001288759.1 | |||
NM_005845.4 | 5494 | UTR 3 | NP_005836.2 | |||
XM_005254025.2 | 5494 | UTR 3 | XP_005254082.1 | |||
XM_017020319.1 | 5494 | UTR 3 | XP_016875808.1 | |||
XM_017020320.1 | 5494 | Intron | XP_016875809.1 | |||
XM_017020321.1 | 5494 | UTR 3 | XP_016875810.1 | |||
XM_017020322.1 | 5494 | Intron | XP_016875811.1 |
Set Membership: |
HapMap |