Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTCCAAAAATGGTTTGCATGTAC[C/T]TATTAATTGAGGTTGACTAAATCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607120 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLCB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PLCB1 - phospholipase C beta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015192.3 | Intron | NP_056007.1 | ||||
NM_182734.2 | Intron | NP_877398.1 | ||||
XM_011529199.2 | Intron | XP_011527501.1 | ||||
XM_011529200.2 | Intron | XP_011527502.1 | ||||
XM_011529201.2 | Intron | XP_011527503.1 | ||||
XM_011529203.2 | Intron | XP_011527505.1 | ||||
XM_017027752.1 | Intron | XP_016883241.1 | ||||
XM_017027753.1 | Intron | XP_016883242.1 |
Set Membership: |
HapMap |