Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGGCTGCTGGGAAGGTTTGCTGC[A/G]ACAAAGGCCTGCATGGCCTACAGAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606157 MIM: 612489 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PANK2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PANK2 - pantothenate kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324191.1 | 1262 | Intron | NP_001311120.1 | |||
NM_001324192.1 | 1262 | Intron | NP_001311121.1 | |||
NM_001324193.1 | 1262 | Intron | NP_001311122.1 | |||
NM_024960.5 | 1262 | Intron | NP_079236.3 | |||
NM_153638.3 | 1262 | Intron | NP_705902.2 | |||
NM_153640.3 | 1262 | Intron | NP_705904.1 |
RNF24 - ring finger protein 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134337.2 | 1262 | UTR 3 | NP_001127809.1 | |||
NM_001134338.2 | 1262 | UTR 3 | NP_001127810.1 | |||
NM_001321749.1 | 1262 | UTR 3 | NP_001308678.1 | |||
NM_007219.4 | 1262 | UTR 3 | NP_009150.1 | |||
XM_011529145.2 | 1262 | UTR 3 | XP_011527447.1 | |||
XM_011529146.2 | 1262 | UTR 3 | XP_011527448.1 | |||
XM_011529147.2 | 1262 | Intron | XP_011527449.1 | |||
XM_017027620.1 | 1262 | UTR 3 | XP_016883109.1 | |||
XM_017027621.1 | 1262 | UTR 3 | XP_016883110.1 | |||
XM_017027622.1 | 1262 | UTR 3 | XP_016883111.1 |
Set Membership: |
HapMap |