Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAGAGTAGAAATGTGAAATCAAGA[A/G]TATTTTTGAGCTACTAAAGCCTTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600192 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SS18 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SS18 - SS18, nBAF chromatin remodeling complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007559.2 | 1557 | UTR 3 | NP_001007560.1 | |||
NM_001308201.1 | 1557 | UTR 3 | NP_001295130.1 | |||
NM_005637.3 | 1557 | UTR 3 | NP_005628.2 | |||
XM_006722527.2 | 1557 | UTR 3 | XP_006722590.1 | |||
XM_011526145.1 | 1557 | UTR 3 | XP_011524447.1 | |||
XM_011526147.2 | 1557 | UTR 3 | XP_011524449.1 | |||
XM_011526148.2 | 1557 | UTR 3 | XP_011524450.1 | |||
XM_011526149.2 | 1557 | UTR 3 | XP_011524451.1 | |||
XM_011526150.2 | 1557 | UTR 3 | XP_011524452.1 | |||
XM_011526151.2 | 1557 | UTR 3 | XP_011524453.1 | |||
XM_011526152.2 | 1557 | UTR 3 | XP_011524454.1 |
Set Membership: |
HapMap |