Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTCCTCATGGCACGTACTCATCA[A/G]TGACAGGTGGTGATTTATATATACA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600529 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AUH PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AUH - AU RNA binding protein/enoyl-CoA hydratase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306190.1 | Intron | NP_001293119.1 | ||||
NM_001698.2 | Intron | NP_001689.1 | ||||
XM_005252066.3 | Intron | XP_005252123.1 | ||||
XM_005252067.4 | Intron | XP_005252124.1 | ||||
XM_005252069.4 | Intron | XP_005252126.1 | ||||
XM_005252072.2 | Intron | XP_005252129.1 | ||||
XM_006717150.3 | Intron | XP_006717213.1 | ||||
XM_011518800.2 | Intron | XP_011517102.1 | ||||
XM_011518802.2 | Intron | XP_011517104.1 | ||||
XM_011518803.2 | Intron | XP_011517105.1 | ||||
XM_011518804.2 | Intron | XP_011517106.1 | ||||
XM_017014849.1 | Intron | XP_016870338.1 | ||||
XM_017014850.1 | Intron | XP_016870339.1 | ||||
XM_017014851.1 | Intron | XP_016870340.1 |
Set Membership: |
HapMap |