Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCACGGGGGATCAAAGTGGAAG[A/G]CTGCGTCCGAATGTACGAACTGGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603371 | ||||||||||||||||||||
Literature Links: |
GLE1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GLE1 - GLE1, RNA export mediator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003722.1 | 615 | Missense Mutation | GAC,GGC | D,G 130 | NP_001003722.1 | |
NM_001499.2 | 615 | Missense Mutation | GAC,GGC | D,G 130 | NP_001490.1 | |
XM_006717059.2 | 615 | Missense Mutation | GAC,GGC | D,G 133 | XP_006717122.1 | |
XM_006717060.2 | 615 | Missense Mutation | GAC,GGC | D,G 133 | XP_006717123.1 | |
XM_011518549.1 | 615 | Missense Mutation | GAC,GGC | D,G 133 | XP_011516851.1 | |
XM_011518550.1 | 615 | Missense Mutation | GAC,GGC | D,G 133 | XP_011516852.1 | |
XM_011518551.1 | 615 | Missense Mutation | GAC,GGC | D,G 130 | XP_011516853.1 |