Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTAGCTGGGAAATGGGAGAATT[A/G]TGCCGAGTGCAAAGCCCGCTCTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607040 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCC11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCC11 - ATP binding cassette subfamily C member 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032583.3 | Intron | NP_115972.2 | ||||
NM_033151.3 | Intron | NP_149163.2 | ||||
NM_145186.2 | Intron | NP_660187.1 | ||||
XM_011523397.1 | Intron | XP_011521699.1 | ||||
XM_011523398.2 | Intron | XP_011521700.1 | ||||
XM_017023795.1 | Intron | XP_016879284.1 | ||||
XM_017023796.1 | Intron | XP_016879285.1 | ||||
XM_017023797.1 | Intron | XP_016879286.1 | ||||
XM_017023798.1 | Intron | XP_016879287.1 | ||||
XM_017023799.1 | Intron | XP_016879288.1 | ||||
XM_017023800.1 | Intron | XP_016879289.1 | ||||
XM_017023801.1 | Intron | XP_016879290.1 | ||||
XM_017023802.1 | Intron | XP_016879291.1 | ||||
XM_017023803.1 | Intron | XP_016879292.1 |