Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATTTTTTTTTTAATGAAGGAAAC[A/G]TTCAATAAGTGAGTCATGAGGTTGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615069 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C12orf60 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C12orf60 - chromosome 12 open reading frame 60 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175874.3 | 1224 | Intron | NP_787070.2 | |||
XM_005253322.4 | 1224 | Intron | XP_005253379.1 | |||
XM_011520568.2 | 1224 | Intron | XP_011518870.1 | |||
XM_011520569.2 | 1224 | Intron | XP_011518871.1 | |||
XM_017018871.1 | 1224 | Intron | XP_016874360.1 | |||
XM_017018872.1 | 1224 | Intron | XP_016874361.1 | |||
XM_017018873.1 | 1224 | Intron | XP_016874362.1 | |||
XM_017018874.1 | 1224 | Intron | XP_016874363.1 |
H2AFJ - H2A histone family member J | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177925.3 | 1224 | UTR 3 | NP_808760.1 |
HIST4H4 - histone cluster 4, H4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105369669 - uncharacterized LOC105369669 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |