Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCGTGCATTCTCCACTGCCGTCT[A/G]CTCGGCCACTCCGGTCCCGACACCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611995 | ||||||||||||||||||||
Literature Links: |
MRPS35 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MRPS35 - mitochondrial ribosomal protein S35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190864.1 | 151 | Missense Mutation | TAC,TGC | Y,C 27 | NP_001177793.1 | |
NM_021821.3 | 151 | Missense Mutation | TAC,TGC | Y,C 27 | NP_068593.2 | |
XM_017019780.1 | 151 | Missense Mutation | TAC,TGC | Y,C 27 | XP_016875269.1 |