Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGATGAACTGGAGCCGTCCTTA[C/T]GGCCTCGGACGCAGGTGAGCTCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610367 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC2A11 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLC2A11 - solute carrier family 2 member 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024938.3 | Intron | NP_001020109.1 | ||||
NM_001024939.3 | Intron | NP_001020110.1 | ||||
NM_001282864.1 | Intron | NP_001269793.1 | ||||
NM_030807.4 | Intron | NP_110434.3 |