Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAAACTTGTTGTTTGTAAAGGAA[A/T]CTGATTTCAGATTAAAATACCTAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
24 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614159 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF644 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF644 - zinc finger protein 644 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016620.3 | 1873 | UTR 3 | NP_057704.2 | |||
NM_032186.4 | 1873 | UTR 3 | NP_115562.3 | |||
NM_201269.2 | 1873 | UTR 3 | NP_958357.1 | |||
XM_005271257.4 | 1873 | UTR 3 | XP_005271314.1 | |||
XM_005271260.4 | 1873 | UTR 3 | XP_005271317.1 | |||
XM_011542258.2 | 1873 | UTR 3 | XP_011540560.1 | |||
XM_011542259.2 | 1873 | UTR 3 | XP_011540561.1 | |||
XM_011542260.2 | 1873 | UTR 3 | XP_011540562.1 | |||
XM_011542261.2 | 1873 | UTR 3 | XP_011540563.1 | |||
XM_017002487.1 | 1873 | UTR 3 | XP_016857976.1 | |||
XM_017002488.1 | 1873 | UTR 3 | XP_016857977.1 | |||
XM_017002489.1 | 1873 | UTR 3 | XP_016857978.1 | |||
XM_017002490.1 | 1873 | UTR 3 | XP_016857979.1 | |||
XM_017002491.1 | 1873 | UTR 3 | XP_016857980.1 | |||
XM_017002492.1 | 1873 | UTR 3 | XP_016857981.1 | |||
XM_017002493.1 | 1873 | UTR 3 | XP_016857982.1 | |||
XM_017002494.1 | 1873 | UTR 3 | XP_016857983.1 |
Set Membership: |
HapMap |