Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGACTACATAGATCAAGGCAGT[G/T]TTCTTCTTTGAGAAACTATCCCAGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120830 MIM: 120831 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C4BPA PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C4BPA - complement component 4 binding protein alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000715.3 | 134 | UTR 5 | NP_000706.1 | |||
XM_005273251.1 | 134 | UTR 5 | XP_005273308.1 | |||
XM_005273252.4 | 134 | UTR 5 | XP_005273309.1 |
C4BPB - complement component 4 binding protein beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |