Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGTCCTTGGGCCCTTCTTCGCCT[C/T]GCCGTCTGTGCAGAATGTGCTAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612196 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABHD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ABHD2 - abhydrolase domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007011.7 | Intron | NP_008942.3 | ||||
NM_152924.4 | Intron | NP_690888.1 | ||||
XM_005254832.1 | Intron | XP_005254889.1 | ||||
XM_005254833.1 | Intron | XP_005254890.1 | ||||
XM_011521165.1 | Intron | XP_011519467.1 |
Set Membership: |
HapMap |