Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGGGAGTTTCTTCCTTCCCTCAG[A/C]TTTCGCCTGGAAGCCTAGAACTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 155760 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACAN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ACAN - aggrecan | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HAPLN3 - hyaluronan and proteoglycan link protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001307952.1 | 1819 | UTR 3 | NP_001294881.1 | |||
NM_178232.3 | 1819 | UTR 3 | NP_839946.1 | |||
XM_011521261.1 | 1819 | UTR 3 | XP_011519563.1 | |||
XM_017021934.1 | 1819 | UTR 3 | XP_016877423.1 | |||
XM_017021935.1 | 1819 | UTR 3 | XP_016877424.1 | |||
XM_017021936.1 | 1819 | UTR 3 | XP_016877425.1 |
Set Membership: |
HapMap |