Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTGGGAGCTGGTCTGAGTGTCCA[C/T]TGAGTCCGTTTATTTGGCGGTCTGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 126350 MIM: 608801 MIM: 600599 MIM: 612256 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNASE2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DNASE2 - deoxyribonuclease II, lysosomal | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GCDH - glutaryl-CoA dehydrogenase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KLF1 - Kruppel like factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011527642.2 | 1458 | UTR 3 | XP_011525944.1 |
MAST1 - microtubule associated serine/threonine kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |