Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCATGACTAATCCGAGATTTTCC[C/T]TGAAATGGAGCCTGAAAGGATCGCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CUEDC1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CUEDC1 - CUE domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271875.1 | Intron | NP_001258804.1 | ||||
NM_001292025.1 | Intron | NP_001278954.1 | ||||
XM_006721900.2 | Intron | XP_006721963.1 | ||||
XM_011524812.2 | Intron | XP_011523114.1 | ||||
XM_017024640.1 | Intron | XP_016880129.1 | ||||
XM_017024641.1 | Intron | XP_016880130.1 |