Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGAAAATTCTAGGTCTAATACA[C/T]GTTAAACAAGTTTCAGGTAATAGTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616102 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMX3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TMX3 - thioredoxin related transmembrane protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019022.3 | 4059 | UTR 3 | NP_061895.3 | |||
XM_005266714.2 | 4059 | UTR 3 | XP_005266771.1 | |||
XM_011526033.2 | 4059 | UTR 3 | XP_011524335.1 | |||
XM_011526034.2 | 4059 | Intron | XP_011524336.1 | |||
XM_011526035.2 | 4059 | Intron | XP_011524337.1 |
Set Membership: |
HapMap |