Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGAATGTATATTTTGATAGGTAG[C/T]TTGAATTTTCTATTTACTCAAACTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 312861 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HTR2C PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HTR2C - 5-hydroxytryptamine receptor 2C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000868.3 | Intron | NP_000859.1 | ||||
NM_001256760.2 | Intron | NP_001243689.1 | ||||
NM_001256761.2 | Intron | NP_001243690.1 |
Set Membership: |
HapMap |