Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCAAAATAAGTATTAAACACCC[A/G]CTATTTGCTTTTTATTATGCAAAGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300195 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMMECR1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AMMECR1 - Alport syndrome, mental retardation, midface hypoplasia and elliptocytosis chromosomal region gene 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025580.1 | Intron | NP_001020751.1 | ||||
NM_001171689.1 | Intron | NP_001165160.1 | ||||
NM_015365.2 | Intron | NP_056180.1 |
SNORD96B - small nucleolar RNA, C/D box 96B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |