Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGTTCCTCAATGTTGTAGAACAT[C/T]CTTGTTTGCCAATAACTGGCAATAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300277 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL1RAPL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IL1RAPL2 - interleukin 1 receptor accessory protein like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017416.1 | Intron | NP_059112.1 | ||||
XM_011530905.2 | Intron | XP_011529207.1 | ||||
XM_017029403.1 | Intron | XP_016884892.1 |
Set Membership: |
HapMap |