Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGACTGTCGGAGAGCGCTTTCCTGA[A/G]AAGTCTGTACTTGTAACGCACCCCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
21 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613126 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYBPHL PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MYBPHL - myosin binding protein H like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010985.2 | 3803 | Intron | NP_001010985.2 | |||
NM_001265613.1 | 3803 | Intron | NP_001252542.1 | |||
XM_017001173.1 | 3803 | Intron | XP_016856662.1 | |||
XM_017001174.1 | 3803 | UTR 3 | XP_016856663.1 | |||
XM_017001175.1 | 3803 | Intron | XP_016856664.1 |
PSRC1 - proline and serine rich coiled-coil 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |