Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTCCTAGTGCTTCTCCTTGACA[C/T]GCACAGGGATGAATGCATTTTCCTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
28 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138380 MIM: 138333 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSTM2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GSTM2 - glutathione S-transferase mu 2 (muscle) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GSTM4 - glutathione S-transferase mu 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000850.4 | Intron | NP_000841.1 | ||||
NM_147148.2 | Intron | NP_671489.1 | ||||
XM_017001085.1 | Intron | XP_016856574.1 | ||||
XM_017001086.1 | Intron | XP_016856575.1 | ||||
XM_017001087.1 | Intron | XP_016856576.1 | ||||
XM_017001088.1 | Intron | XP_016856577.1 |
Set Membership: |
HapMap |