Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCCAAAGTCAGGTCTGTAGCAGA[C/G]TCCGGCCAGCTGACTGGAGACGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138385 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSTM5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GSTM5 - glutathione S-transferase mu 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000851.3 | Intron | NP_000842.2 | ||||
XM_005270784.4 | Intron | XP_005270841.1 | ||||
XM_005270785.4 | Intron | XP_005270842.1 |
Set Membership: |
HapMap |