Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAAGCTGGCAGCTGGGGTTCTTCC[C/T]GAGTCCACTCCCCTGATCCTTGCTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
22 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614989 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPS8L3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
EPS8L3 - EPS8 like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319952.1 | Intron | NP_001306881.1 | ||||
NM_024526.3 | Intron | NP_078802.2 | ||||
NM_133181.3 | Intron | NP_573444.2 | ||||
NM_139053.2 | Intron | NP_620641.1 | ||||
XM_011542132.1 | Intron | XP_011540434.1 | ||||
XM_011542133.1 | Intron | XP_011540435.1 | ||||
XM_011542134.2 | Intron | XP_011540436.1 | ||||
XM_011542135.2 | Intron | XP_011540437.1 | ||||
XM_017002327.1 | Intron | XP_016857816.1 | ||||
XM_017002328.1 | Intron | XP_016857817.1 | ||||
XM_017002329.1 | Intron | XP_016857818.1 |
Set Membership: |
HapMap |