Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTCCATCTGTGTCTTTCTGGTC[C/T]GGAGATTCTGCTGCTTTGGTAACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600682 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKR7A2P1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AKR7A2P1 - aldo-keto reductase family 7 member A2 pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC16A1 - solute carrier family 16 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166496.1 | 1815 | Silent Mutation | CCA,CCG | P,P 484 | NP_001159968.1 | |
NM_003051.3 | 1815 | Silent Mutation | CCA,CCG | P,P 484 | NP_003042.3 | |
XM_011542026.2 | 1815 | Intron | XP_011540328.1 | |||
XM_011542027.2 | 1815 | Intron | XP_011540329.1 |
Set Membership: |
DME Validated Inventoried |